Learn More
Medchemexpress LLC Geldanamycin | 30562-34-6 | C₂₉H₄₀N₂O₉ | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1523010MM/1ML
Encompass_Preferred
Geldanamycin is an Hsp90 inhibitor exhibiting antimicrobial activity against numerous Gram-positive and some Gram-negative bacteria. It also shows anti-influenza virus H5N1 activities. This natural product inhibitor binds to the N-terminal ATPase domain of Hsp90, inhibiting its chaperone function and inducing tumor cell death via apoptosis. It protects against neuronal injury and decreases RIP1 protein levels.
  • Hsp90 inhibitor
  • Exhibits antimicrobial activity
  • Shows anti-influenza virus H5N1 activities
  • Protects against neuronal injury
  • Decreases RIP1 protein levels
  • Induces tumor cell death via apoptosis
Catalog No. 50-003-73884