Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Geldanamycin-FITC | 2969156-01-0 | MFCD32068377 | 98.4% | 1,034.18 g·mol⁻¹ | C55H63N5O13S | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1337055MG

Encompass_Preferred

Geldanamycin-FITC is a fluorescently labeled geldanamycin derivative used as a probe for Hsp90-binding studies, fluorescence polarization assays, and cellular imaging. It is supplied as a light brown solid of high purity intended for research applications requiring a fluorescent Hsp90 ligand.

  • High purity (98.43%) for reliable assay results.
  • Emission at ~515 nm suitable for fluorescence detection.
  • Soluble in DMSO at 25 mg/mL; may require ultrasonication.
  • Light brown solid; store protected from light and at low temperature.
  • Suitable for Hsp90 binding assays, fluorescence polarization, and cellular imaging.

Catalog No. 50-002-62927


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.