Learn More
Abcam Geldanamycin, Hsp90 inhibitor, 1MG
Supplier:  Abcam AB1208961MG
Encompass_Preferred
MW 560.64 g/mol, Purity >98%. Selective Hsp90 inhibitor. Binds the ATP site of Hsp90 and inhibits chaperone activity. A benzoquinone ansamycin antibiotic. Also inhibits the Hsp90 paralog GRP94. Exhibits various cellular actions including gene expression, cell proliferation, apoptosis and angiogenesis.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7155