Learn More
Medchemexpress LLC Geldanamycin (Standard) | 30562-34-6 | C29H40N2O9 | 25 MG

Supplier: Medchemexpress LLC 25MGHY15230
Geldanamycin is an Hsp90 inhibitor with antimicrobial activity against various Gram-positive and some Gram-negative bacteria, and also demonstrates anti-influenza virus H5N1 activities. This analytical standard protects against neuronal injury and decreases RIP1 protein levels. It is recognized as the first natural product inhibitor of Hsp90, binding to its N-terminal ATPase domain to inhibit chaperone function and induce tumor cell death through an apoptotic mechanism.
- Hsp90 inhibitor
- Antimicrobial activity
- Anti-influenza virus H5N1 activities
- Protects against neuronal injury
- Decreases RIP1 protein levels
- Purity 99.43%
- Molecular weight 560.64
- Light yellow to orange solid
- Initial source: Microorganisms (Streptomyces hygroscopicus)
- Kd for Hsp90: 1.2 μM
- Solubility in DMSO: 25 mg/mL
- Melting/freezing point: 255°C
- Stable under recommended storage conditions
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.