Learn More
Medchemexpress LLC GelHP Protein Gel (Tris-Gly, 15%, 15 well) | 10 EA
SDP
Supplier:  Medchemexpress LLC 10PCSHYK1116
Encompass_Preferred
This GelHP protein gel is a precast polyacrylamide gel, offering a safe, convenient, and high-performance solution for protein separation. It is suitable for both PAGE and Western blot analysis, formulated without SDS for use in both denaturing and native electrophoresis.
  • Coated plastic gel plates minimize non-specific protein adsorption for sharper bands.
  • Easy-open cassette design for effortless gel plate removal.
  • SDS-free formulation compatible with denaturing and native PAGE.
  • Automated gel casting ensures high batch-to-batch stability and reproducibility.
  • Compatible with most mainstream mini gel electrophoresis systems.
  • Available in multiple uniform and gradient concentrations.
Catalog No. 50-003-65394