Learn More
Medchemexpress LLC Gellan gum | 71010-52-1 | 97.0% | Unspecified (polymeric polysaccharide; no fixed molecular formula) | 100MG
SDP
Supplier:  Medchemexpress LLC HY125870100MG
Encompass_Preferred
Gellan gum is a high-molecular-weight microbial exopolysaccharide used as an agar substitute and gelling agent for cell culture. It forms firm, transparent gels at low concentrations and can act as a scaffold for three-dimensional cell culture and tissue engineering. Supplied as a white to off-white powder in small research pack sizes.
Catalog No. 50-002-35325