Learn More
Medchemexpress LLC GelMA, 30% methacrylation | 2280018-77-9 | 30% | 10 MG
SDP
Supplier:  Medchemexpress LLC 10MGHYW763582
Encompass_Preferred
GelMA (Gelatin Methacryloyl), 30% methacrylation, is a derivative obtained by the reaction of Methacrylic anhydride (MA) and gelatin. It exhibits excellent biocompatibility, biodegradability, and moldability. GelMA can be photocrosslinked into hydrogels.
  • Used for research in tissue regeneration
  • Suitable for drug delivery
  • Applied in organ-on-a-chip research
  • Used for biosensing
Catalog No. 50-003-65231