Learn More
Cayman Chemical Gelsevrne 5mg
SDP
Supplier:  Cayman Chemical 397495 MG
Encompass_Preferred
An oxindole alkaloid with anti-inflammatory activity; binds to STING (Kd = 27.6 µM) and prevents 2'3'-cGAMP-induced Sting dimerization in RAW 264.7 cells at 5 µM; inhibits 2'3'-cGAMP-, poly(dA:dT) dsDNA-, or IFN-stimulatory DNA-induced increases in the levels of mRNA encoding CXCL10 and IL-6 in THP-1 monocytes and RAW 264.7 cells at 10 µM; inhibits 2'3'-cGAMP-induced increases in the levels of Sting and phosphorylated IRF3, p65, and TBK1 in RAW 264.7 cells at 5 µM; increases animal survival time, reduces lung injury and total protein levels in BALF, and decreases serum levels of ALT, AST, creatinine, IL-6, TNF-α, and BUN levels in a mouse model of sepsis induced by CLP at 20 mg/kg
Catalog No. 50-294-3028