Learn More
Medchemexpress LLC HY-17026 200mg , Gemcitabine CAS:95058-81-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY17026200MG
Encompass_Preferred
HY-17026 200mg Gemcitabine CAS: 95058-81-4 Gemcitabine (LY 188011) is a pyrimidine nucleoside analog antimetabolite and an antineoplastic agent. Gemcitabine inhibits DNA synthesis and repair, resulting in autophagyand apoptosis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9264