Learn More
Selleck Chemical LLC Gemcitabine S1714-1g
SDP
Supplier:  Selleck Chemical LLC S17141G
Encompass_Preferred
Gemcitabine a nucleic acid synthesis inhibitor is a very potent and specific deoxycytidine analogue used as chemotherapy Gemcitabine induces a potent p53-dependent apoptosis
Catalog No. 50-311-0655