Learn More
Medchemexpress LLC Gemcitabine | 95058-81-4 | 263.20 | 5 G
SDP
Supplier:  Medchemexpress LLC HY170265G
Encompass_Preferred
Gemcitabine (LY 188011) is a pyrimidine nucleoside analog antimetabolite and an antineoplastic agent. It inhibits DNA synthesis and repair, and can modulate autophagy. Gemcitabine induces apoptosis through the activation of p38 MAPK. It has demonstrated efficacy in mouse models of pancreatic and breast cancer and can be used for cancer research, including pancreatic cancer, non-small cell lung cancer, and breast cancer.
  • Inhibits DNA synthesis and repair
  • Modulates autophagy
  • Induces apoptosis through p38 MAPK activation
  • Demonstrates efficacy in mouse models of pancreatic and breast cancer
  • Suitable for research in pancreatic cancer, non-small cell lung cancer, and breast cancer
Catalog No. 50-003-84207