Learn More
Selleck Chemical LLC Gemcitabine HCl 100mg 122111-03-9 LY188011
SDP
Supplier:  Selleck Chemical LLC S1149100MG
Encompass_Preferred
Gemcitabine HCl (LY188011) is a DNA synthesis inhibitor with IC50 of 50 nM, 40 nM, 18 nM and 12 nM in PANC1, MIAPaCa2, BxPC3 and Capan2 cells, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-673-1