Learn More
Medchemexpress LLC Gemcitabine hydrochloride | 122111-03-9 | 99.9% | 299.66 | 100 MG
SDP
Supplier:  Medchemexpress LLC 100MGHYB0003
Encompass_Preferred
Gemcitabine Hydrochloride (LY 188011 Hydrochloride) is a pyrimidine nucleoside analog antimetabolite and an antineoplastic agent. It inhibits DNA synthesis and repair, leading to autophagy and apoptosis.
  • Acts as an antineoplastic agent
  • Inhibits DNA synthesis and repair
  • Induces autophagy and apoptosis
  • Targets DNA/RNA synthesis
  • Functions as a nucleoside antimetabolite/analog
  • Used in cancer research
Catalog No. 50-003-65018