Learn More
Medchemexpress LLC Gemcitabine hydrochloride (Standard) | 122111-03-9 | 99.9% | C9H12ClF2N3O4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0003R100MG
Encompass_Preferred
Gemcitabine hydrochloride (Standard) is a pyrimidine nucleoside analog antimetabolite and an antineoplastic agent. It inhibits DNA synthesis and repair, leading to autophagy and apoptosis. This product is typically used as an analytical standard.
  • Inhibits DNA synthesis and repair
  • Induces autophagy and apoptosis
  • Used as an analytical standard
  • High purity for accurate results
Catalog No. 50-004-32398