Learn More
Medchemexpress LLC Gemcitabine hydrochloride (Standard) | 122111-03-9 | 99.94% | 299.66 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY17026R
Encompass_Preferred
Gemcitabine hydrochloride (Standard) serves as the analytical standard for Gemcitabine (hydrochloride), primarily for research and analytical applications. This product, also known as LY 188011 Hydrochloride, is a pyrimidine nucleoside analog antimetabolite and an antineoplastic agent that functions by inhibiting DNA synthesis and repair, ultimately inducing autophagy and apoptosis.
  • Analytical standard for research and analytical applications.
  • Functions as a pyrimidine nucleoside analog antimetabolite.
  • Acts as an antineoplastic agent.
  • Inhibits DNA synthesis and repair.
  • Induces autophagy and apoptosis.
  • Utilized in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
Catalog No. 50-003-85320