Learn More
Medchemexpress LLC Gemcitabine monophosphate | 116371-67-6 | 99.98% | 343.18 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1089321MG
Encompass_Preferred
Gemcitabine monophosphate is an active intermediate of Gemcitabine. It demonstrates a synergistic anti-cancer effect and can be delivered through nanoparticle formulation. It is intended for research use only.
  • Active intermediate of Gemcitabine.
  • Exhibits synergistic anti-cancer effects.
  • Can be delivered via nanoparticles.
  • Induces tumor cell apoptosis (30-40%).
  • Reduces tumor cell proliferation (8 times).
  • Significantly reduces tumor microvessel density (MVD).
  • Inhibits tumor mitosis.
Catalog No. 50-003-45310