Learn More
Medchemexpress LLC Gemcitabine monophosphate | 116371-67-6 | 99.98% | 343.18 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY108932
Encompass_Preferred
Gemcitabine monophosphate (Gemcitabine 5′-phosphate) is an active intermediate of Gemcitabine. It exhibits a synergistic anti-cancer effect and can be delivered through formulation into nanoparticles. It is intended for research use only. The product appears as a solid, white to off-white in color.
  • Functions as an active intermediate.
  • Demonstrates a synergistic anti-cancer effect.
  • Can be effectively delivered by formulating it into nanoparticles.
  • An in vivo study showed that combined treatment exhibited anti-small cell lung cancer activity and induced apoptosis.
  • Soluble in DMSO (200 mg/mL) and H2O (100 mg/mL).
Catalog No. 50-003-47272