Learn More
Medchemexpress LLC Gemcitabine-O-Si(di-iso)-O-Mc | 98.82% | 558.65 | 10 MG
SDP
Supplier:  Medchemexpress LLC 10MGHY130812
Encompass_Preferred
Gemcitabine-O-Si(di-iso)-O-Mc is an agent-linker conjugate for ADC with potent antitumor activity. It uses Gemcitabine (a pyrimidine nucleoside analog antimetabolite and an antineoplastic agent), linked via an ADC linker. Antibody drug conjugates (ADC) couple therapeutic monoclonal antibodies (mAb) with potent toxins through a linker that is stable within systemic circulation but cleaves within the target cells.
  • Drug-Linker conjugates for ADC
  • Cleavable linker
  • Antibody-drug conjugates
  • Pyrimidine nucleoside analog antimetabolite
  • Antineoplastic agent
  • Inhibitor
  • Cancer targeted therapy
  • Cancer immunotherapy
Catalog No. 50-003-65298