Learn More
Medchemexpress LLC Gemcitabine (standard) | 95058-81-4 | 99.99% | 263.20 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY17026R100MG
Encompass_Preferred
Gemcitabine (Standard) is an analytical standard intended for research and analytical applications. It is a pyrimidine nucleoside analog antimetabolite and an antineoplastic agent that inhibits DNA synthesis and repair, leading to autophagy and apoptosis.
  • Analytical standard for assay reference
  • Used in qualitative, quantitative, and methodological research experiments
  • Applicable in HPLC, GC, and MS techniques
  • Inhibits DNA synthesis and repair
  • Induces autophagy and apoptosis
Catalog No. 50-003-84496