Learn More
Medchemexpress LLC Gemcitabine (Standard) | 95058-81-4 | 99.99% | 263.20 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY17026R5MG
Encompass_Preferred
Gemcitabine (Standard) is an analytical standard of Gemcitabine, used in research and analytical applications. This pyrimidine nucleoside analog acts as an antimetabolite and antineoplastic agent by inhibiting DNA synthesis and repair, which leads to autophagy and apoptosis. It serves as a reference standard for assays and is commonly employed in qualitative, quantitative, and methodological research experiments.
  • Inhibits DNA synthesis and repair
  • Leads to autophagy and apoptosis
  • Analytical standard grade
  • Reference standard for assays
  • Used in HPLC, GC, and MS experiments
Catalog No. 50-003-84140