Learn More
Medchemexpress LLC Gemcitabine triphosphate trisodium | 110988-86-8 | 99.7% | 569.08 g·mol⁻¹ | C9H11F2N3Na3O13P3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY17026E10MG
Encompass_Preferred
Gemcitabine triphosphate trisodium is the triphosphate metabolite of the nucleoside analog gemcitabine, provided as a research-grade standard for biochemical and cellular studies. Supplied as a high-purity trisodium salt, it is intended for use as an analytical standard in metabolism assays, uptake and retention studies, and imaging or radiolabeling experiments.
  • High purity (99.7%) suitable for analytical applications.
  • Supplied as a trisodium salt with molecular weight 569.08 g·mol⁻¹.
  • Liquid, colorless to light yellow appearance for easy handling.
  • Requires low-temperature storage at -80°C to maintain stability.
  • Useful as a standard for assays measuring intracellular triphosphate levels.
  • Applicable for radiolabeled probe imaging and uptake studies.
Catalog No. 50-002-95129