Learn More
Cayman Chemical GemIfloxacInmesylate 5mg
SDP
Supplier:  Cayman Chemical 210475 MG
Encompass_Preferred
A fluoroquinolone antibiotic that is effective against C. pneumoniae and M. tuberculosis (MIC50s = 0.25 and 8 UG/ml, respectively); useful against respiratory tract infections
Catalog No. 50-298-4171