Learn More
Phenomenex Inc Phenomenex LC Column, NX-C18, 5 μm, 250 x 4.6 mm
Supplier:  Phenomenex Inc 00G4454EO
Encompass
The Phenomenex Gemini NX-C18 reversed-phase HPLC column delivers robust C18 selectivity, high efficiency, and extended pH stability for demanding analytical and preparative separations.
  • 5 μm particle size for high separation efficiency.
  • 110 Å pore size for small to mid-size molecules.
  • 250 x 4.6 mm analytical column dimensions.
  • Endcapped C18 phase for improved peak shape and reproducibility.
  • Approximate carbon load 14% for strong retention characteristics.
  • Isocratic pH range 1-12 for extreme pH applications.
  • Suitable for high-efficiency analytical or preparative separations.
Catalog No. NC3713215