Learn More
DIATRON US Gemini Automated ELISA Immunoassay System
Supplier:  DIATRON US 9162800000
Encompass_Preferred
Gemini is an Automated 96 Well ELISA System are computer-controlled microplate processing systems that fully automates ELISA assays. The Gemini line automates the sample distribution, incubation, reagent addition, washing, and detection phases of microplate assays.
Catalog No. NC1847131
Add to Cart
Request a Quote