Learn More
Sigma Aldrich Fine Chemicals Biosciences Gemcitabine hydrochloride | 122111-03-9 | MFCD01735988 | 10 mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences G642310MG
Encompass
Gemcitabine hydrochloride | Purity: ≥98% | Mol Wt: 299.66 | 122111-03-9 | MFCD01735988 | 10 mg
Catalog No. NC0824938