Learn More
Grainger Commercial Ice Machines
Supplier:  Grainger 36RF31
Encompass_Preferred
The website offers a range of commercial ice machines suitable for restaurants, bars, hotels, and stores. Models vary from undercounter to freestanding, producing 50-1500 lbs of ice daily. Features include stainless steel, air or water-cooled units, automatic shutoff, and self-diagnostics. Brands like Manitowoc and Scotsman are available, ensuring businesses can efficiently meet their ice needs for beverages and food preservation.
Catalog No. NC2872300
Only null left
Add to Cart
Request a Quote