Learn More
MALIX LLC OTC Covid Home Test: Conveniently check for Covid-19 from the comfort of your home with this OTC at-home self-test. It uses a non-invasive short nasal swab and delivers results in just 15 minutes. For quick and easy results.
Supplier:  MALIX LLC GENABIO1PK
Encompass_Preferred
Genabio COVID-19 Test 1 Pack
OTC Covid Home Test: Conveniently check for Covid-19 from the comfort of your home with this OTC at-home self-test. It uses a non-invasive short nasal swab and delivers results in just 15 minutes, providing quick and easy results.Trusted by Professionals: The Genabio COVID-19 Rapid Self-Test Kit has been authorized by the FDA for emergency use and is used by healthcare professionals across the USA.Home Convenience: Avoid the hassle of scheduling an appointment and waiting in line at a testing center by using the Genabio COVID-19 Rapid Self-Test Kit in the comfort of your own home.Quick and Easy: With the attached buffer fluid lid, this test is quick and easy to complete, reducing the risk of spills compared to other brands.buffer fluid lid it makes for a quick and easy test that can be completed faster and without spills compared to other brands..
Catalog No. NC3235794