Learn More
Epoch Life Science Inc EconoSpin® All-In-One DNA/RNA Mini Spin Columns w/o lid (250 columns with collection tubes)
SDP
Supplier:  Epoch Life Science Inc 1910250
Encompass_Preferred
Silica membrane mini spin columns with collection tubes for gel PCR & plasmid purification. Sample volume: 850 ul. Binding capacity: 45 ug plasmid DNA. Minimum elution volume: 20 ul. Compatible with third party buffer sets. This item does not include a lid. 18 month shelf life. Used by labs around the world.
Catalog No. NC0231536