Learn More
Genscript Corporation Antibody Enzyme Other Products Molecular Biology 1 MG 1/EA
Supplier:  Genscript Corporation Z036221
Encompass
GenCRISPR Ultra eSpCas9-2NLS is a mutant form of Cas9 nuclease and is produced by expression in an E. coli strain carrying a plasmid encoding the eSpCas9 gene from Streptococcus pyogenes with double-ended nuclear localization signal (NLS). GenCRISPR Ultra eSpCas9-2NLS delivers higher fidelity and less off-target activity than wild-type SpCas9 nuclease.
Catalog No. 50-252-9587