Learn More
Sigma Aldrich Fine Chemicals Biosciences GenEluteTM Plasmid Minipre
Supplier:  Sigma Aldrich Fine Chemicals Biosciences PLN701KT
Encompass
We are committed to bringing you Greener Alternative Products which adhere to one or more of The 12 Principles of Greener Chemistry. This product has Inherently Safer Chemistry compared to the standard use of phenol and chloroform to perform DNA extractions.
Catalog No. 50-290-1599