Learn More
GA International Thermal-transfer, smudge and alcohol-proof resin ribbons 2.24" x 243' / 57mm x 74m Black 0.5" / 12.7mm
Supplier:  GA International RR57X74C0.51IZ2
Encompass_Preferred
Smudge and alcohol proof resin ribbon for thermal-transfer printers. Available in either black or white ink formats, the printout obtained when printing with this ribbon will resist spraying and rubbing with alcohols. Range of sizes available, ensuring compatibility with most common barcode printer brands.
Catalog No. NC3706096