Learn More
Medchemexpress LLC Genevant CL1 | 1450888-71-7 | >99.9% | C39H73NO2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY467595MG
Encompass_Preferred
Genevant CL1 is an ionizable lipid with a pKa of 6.3, utilized in the formulation of lipid nanoparticles (LNPs) for the efficient delivery of nucleic acid payloads. It plays a crucial role in intramuscular vaccines and is a key component in LNP delivery systems for mRNA vaccines.
  • Ionizable lipid with a pKa of 6.3
  • Suitable for preparing lipid nanoparticles (LNPs)
  • Delivers nucleic acid payloads
  • Used in intramuscular vaccines
  • Component of LNP delivery systems for mRNA vaccines
Catalog No. 50-003-58464