Learn More
Medchemexpress LLC Genistein | 446-72-0 | MFCD00016952 | 99.3% | 270.24 g/mol | C15H10O5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY14596R5MG
Encompass_Preferred
Genistein (Standard) is an analytical reference standard intended for research and analytical applications, used for calibration and method validation in chromatographic and mass-spectrometry assays. Genistein is a soy isoflavone studied as a tyrosine kinase inhibitor with reported effects on apoptosis, the cell cycle, angiogenesis, and metastasis.
  • High purity (99.3%).
  • Suitable as a calibration and reference material for HPLC and LC-MS.
  • Available in small quantities for analytical use (e.g., 5 mg).
  • Consistent molecular identity: C15H10O5, 270.24 g/mol.
  • Intended for laboratory research and analytical applications.
Catalog No. 50-002-22283