Learn More
Medchemexpress LLC HY-14596 500mg , Genistein NPI 031L CAS:446-72-0 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY14596500MG
Encompass_Preferred
Medchemexpress, HY-14596 500mg Genistein NPI 031L CAS:446-72-0 Formula:C15H10O5 IC50: 0.7 μg/mL (0.6 μM) (EGFR) Purity:98% Genistein is a potent inhibitor of the protein tyrosine kinase (PTK) activity of the EGFR in vitro with an IC 50 of 0.7 μg/mL (0.6 μM). Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2365
 Restricted Item