Learn More
Cayman Chemical GenIsteIn-d4 500ug
SDP
Supplier:  Cayman Chemical 18244500 UG
Encompass_Preferred
An internal standard for the quantification of genistein by GC- or LC-MS
Catalog No. 50-296-6039