Learn More
Selleck Chemical LLC Genistin (Genistoside) S2301-50mg
SDP
Supplier:  Selleck Chemical LLC S230150MG
Encompass_Preferred
Genistin (Genistoside Genistine Genistein 7-glucoside Genistein glucoside) is an isoflavone found in a number of dietary plants like soy and kudzu Genistin attenuates cellular growth and promotes apoptotic cell death breast cancer cells through modulation of Estrogen Receptor (ER ) signaling pathway
Catalog No. 50-311-2648