Learn More
Zymo Research Corporation Genomic DNA Clean & Concentrator™ Kit-5 (2 x 96 Preps)
SDP
Supplier:  Zymo Research Corporation D4066
Encompass_Preferred
There is no need for organic denaturants, chloroform, or messy precipitations: simply add the specially formulated ChIP DNA Binding Buffer to a sample and then transfer the mixture to the supplied ZymoSpin™ I-96-XL Plate. Eluted DNA is suitable for sequencing, PCR, endonuclease digestion, and other enzymatic procedures. The product is also compatible with smaller DNAs (50 bp to 10 kb) from PCR, digestions, crude plasmid preparations, cDNA synthesis, etc.
Catalog No. 50-125-1423