Learn More
ATCC ATCC Genomic DNA from Acinetobacter baumannii, 5 μg, BAA-1605D-5
Supplier: ATCC BAA1605D5
Purified genomic DNA isolated from Acinetobacter baumannii (Bouvet and Grimont). Supplied as 5 μg of dried DNA in 1X TE buffer, this genomic DNA is intended for molecular biology workflows including PCR, whole-genome sequencing, and assay development.
- Organism: Acinetobacter baumannii (Bouvet and Grimont).
- Product type: Genomic DNA (nucleic acid).
- Quantity: 5 μg.
- Format: Dried.
- Storage buffer: 1X TE buffer.
- Genome sequenced: Yes.
- Applications: PCR, sequencing, assay development.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.