Learn More
LUNA NANOTECH INC Genomic Dna Purification Kit, 96 Well Plate Format, 384 Preps
Supplier:  LUNA NANOTECH INC NKP061384
Encompass_Preferred
PuroSPIN-96™ Genomic DNA Purification Kit, 96-Well Plate Format, 384 Preps
The PuroSPIN-96™ Genomic DNA Purification Kit is designed to purify genomic DNA from different biological samples such as bacteria, yeast, mammalian cells, blood, and animal tissue using 96-well silica membrane purification plates. The genomic DNA is released from the cells or tissue using Lysis Buffer  LB-GD1, Lysis Buffer LB-GD2, and Proteinase K. The DNA is then loaded onto silica-based purification plates and washed to remove protein, salt, and other contaminants. Finally, the purified DNA is eluted from the column into a clean 96-well elution plate in Elution Buffer EB, or nuclease-free water.
High quality DNA extracted with the PuroSPIN™ 96 Genomic DNA Purification Kit can be used for a range of downstream applications such as PCR, Southern blotting, or genomic sequencing.
 Features:
- Easily purify genomic DNA from a va
Catalog No. 50-256-6921