Learn More
ATCC Genomic RNA from; SARS-CoV-2; Delta variant; Strain: USA/MD-HP05285/2021
Supplier:  ATCC VR3342D
Encompass
Genomic RNA isolated from a preparation of Severe acute respiratory syndrome-related coronavirus 2 isolate USA/MD-HP05285/2021. This product can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine viral load.
Catalog No. 50-238-2378
 Restricted Item