Learn More
MSC CRC Lithium General Purpose Grease, NLGI Grade 2, 14 oz
Supplier:  MSC 48738066
Encompass
Sta-Lube Lithium General Purpose Grease is an NLGI Grade 2, lithium-based lubricant that is water-resistant and extremely stable. Designed for light-duty applications, it combines high-viscosity grease with the adaptability of liquid oil, offering excellent water resistance and superior metal protection with rust and oxidation inhibitors.
  • Excellent water resistance
  • Extremely stable for light-duty applications
  • Contains rust and oxidation inhibitors for superior metal protection
  • Typical drop point: 350°F
  • Will not break down or run out
  • Offers superior durability under environmental stresses
  • Reduces friction and wear on bearings
  • Stays in place once applied
Catalog No. 50-224-1093