Learn More
Medchemexpress LLC Gentamicin | 1403-66-3 | C24H53N7O73* | 5 MG
SDP
Supplier:  Medchemexpress LLC HYA0276A5MG
Encompass_Preferred
Gentamicin is an aminoglycoside antibiotic that inhibits the growth of both gram-positive and gram-negative bacteria, and it also inhibits several strains of mycoplasma in tissue culture. It inhibits DNase I with an IC50 of 0.57 mM.
  • More effective in vitro bacterial inhibitor than combined penicillin-streptomycin
  • Nontoxic to tissue culture monolayers
  • Does not inhibit virus replication
  • Has been successfully used as an additive in commercial mycology media to inhibit bacterial growth
  • Bactericidal for a wider range of organisms (Pseudomonas aeruginosa, Proteus sp., and Streptococcus faecalis) compared to penicillin and streptomycin
  • Does not interfere with the production of cytopathic effect by certain echoviruses and polioviruses in tissue culture
  • Nontoxic to Rhesus monkey kidney, HeLa, and human amnion cells
  • Stable at autoclave temperatures
Catalog No. 50-004-32948