Learn More
Quality Biological Inc Gentamicin 50 mg/mL, PK 5 X 10 mL
SDP
Supplier:  Quality Biological Inc 120098661
Encompass_Preferred
Gentamicin is an aminoglycoside complex produced by fermentation of Micromonospora purpurea or M. echinospora. ¿ It is used as the sulfate salt. ¿ Ready to use solution ¿ Applications: Gentamicin sulfate is a broad-spectrum antibiotic used as a selection agent (gentamicin-resistance gene) in molecular biology and cell culture applications. ¿ This product is formulated to contain 50 mg/mL Gentamicin in deionized water ¿ Recommended for use in eukaryotic cell culture at a volume of 1 milliliter per liter. ¿ Storage Conditions: 15¿C to 30 ¿C ¿ Shipping Condition: ambient temperature ¿ Sterile Filtered ¿ Product and/or packaging contains recycled/renewable content ¿ Product and/or packaging is recyclable or reduces waste ¿ Product and/or packaging is safer to human and environmental health ¿ UNSPSC: 41122100 ¿ ¿ Max Shelf Life: 36 months
Catalog No. 50-983-230