Learn More
PHYTOTECHNOLOGY LABS GENTAMICIN SOLUTION 100ML
Supplier:  PHYTOTECHNOLOGY LABS G3410100ML
Encompass_Preferred
Gentamicin Solution (100mg/ml)Gentamicin sulfate is an aminoglycoside antibiotic that is effective against many gram-negative bacteria (e.g., Brucella, Escherichia, Enterobacter, Francisella, Yersinia, etc.) and some strains of staphylococci.
Catalog No. NC1789708