Learn More
Alkali Scientific Gentamicin Sulfate, 1 G
Supplier: Alkali Scientific GEN001CP
Gentamicin Sulfate (1g) is an aminoglycoside antibiotic commonly used to treat various bacterial infections. It works by inhibiting bacterial protein synthesis, leading to the death of susceptible bacteria. Gentamicin is effective against both Gram-positive and Gram-negative bacteria, and it is frequently used for treating infections such as pneumonia, sepsis, and urinary tract infections. This 1g powder form is ideal for preparation in laboratory settings and clinical use. It is particularly valuable for infections where resistance to other antibiotics is present. Gentamicin is often used in combination with other antibiotics for synergistic effects in severe infections.