Learn More
Alkali Scientific Gentamicin Sulfate, 50 G
SDP
Supplier:  Alkali Scientific GEN005CP
Encompass_Preferred
Gentamicin Sulfate (50g) is a highly effective aminoglycoside antibiotic used for treating a broad range of bacterial infections. It works by binding to bacterial ribosomes, inhibiting protein synthesis, and preventing bacterial growth. This 50g powder formulation is prepared for antibacterial use and is ideal for preparing large volumes of solution for both research and clinical applications. Gentamicin is particularly effective against Gram-negative bacteria, including Pseudomonas and Escherichia coli. This large quantity is useful for laboratories that require significant amounts for experimental work or clinical use, particularly in treating serious infections or in settings where multidrug-resistant bacteria are prevalent.
Catalog No. 50-006-61887