Learn More
FUJIFILM BIOSCIENCES INC Gentamicin Sulfate Solution
Supplier:  FUJIFILM BIOSCIENCES INC 935520ML
Encompass
Inhibits the growth of a wide variety of Gram-positive and Gram-negative microorganisms Also effective against various strains of mycoplasma Recommended concentration for cell culture application 1ML/L medium
Catalog No. 50-004-50290