Learn More
Medchemexpress LLC Gentisuric acid | 25351-24-0 | MFCD00059630 | 99.7% | 211.17 g/mol | C9H9NO5 | 1 ML

Supplier: Medchemexpress LLC HYW33885210MM/1ML
Gentisuric acid is a biochemical research reagent and an aspirin metabolite used in studies of peptidylglycine alpha-amidating monooxygenase (PAM) activity and DNA damage pathways. It has been reported to prevent Mitomycin C-induced DNA damage and is provided in a solution format suitable for in vitro assays.
- Used as a reagent for studying aspirin metabolism and PAM activity.
- Reported to prevent Mitomycin C-induced DNA damage in experimental studies.
- Purity approximately 99.65%.
- CAS number 25351-24-0.
- Molecular formula C9H9NO5; molecular weight 211.17 g/mol.
- Supplied as a 10 mM solution, 1 mL (solvent not specified).
- Suitable for in vitro biochemical and cellular assays; follow SDS for handling and storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.