Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Gentisuric acid | 25351-24-0 | MFCD00059630 | 99.7% | 211.17 g/mol | C9H9NO5 | 250 MG
SDP

Supplier:  Medchemexpress LLC HYW338852250MG

Encompass_Preferred

Gentisuric acid is provided as an analytical standard of the aspirin metabolite for research and analytical applications. It is reported as a substrate of peptidylglycine alpha-amidating monooxygenase (PAM) and has been described to reduce mitomycin C-induced DNA damage in published studies. Typical uses include analytical reference, metabolic profiling, and method development.

  • Analytical standard for aspirin metabolite studies.
  • Reported purity 99.7%.
  • Molecular formula C9H9NO5.
  • Molecular weight 211.17 g/mol.
  • Typical pack size 250 mg.
  • Intended for research and analytical applications.

Catalog No. 50-002-82095


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.