Learn More
Enterprise Technology Solutions Genuine Joe Anti-fatigue Mat, 3/8" Vinyl Foam, 2'x3', Gray
SDP
Supplier:  Enterprise Technology Solutions GJO00085
Encompass_Preferred
Anti-fatigue mat offers 3/8" thick, vinyl foam structure to provide support that reduces fatigue and discomfort. Ribbed surface increases safety. Beveled edge allows easy wheel movement on and off the mat. Mat is designed for indoor use in warehouses, factories and more.
Catalog No. 50-008-10901